Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(2-Naphthyl)-DL-alanine, 98%

Chemical Identifiers

Chemical Name or Material 3-(2-Naphthyl)-DL-alanine
CAS 14108-60-2
Formula Weight 215.25
MDL Number MFCD00070216
Synonym DL-Ala(2-naphthyl)-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6648703

  SDS
alfa aesar™
H6648703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6648706

  SDS
alfa aesar™
H6648706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH66487MD

  SDS
alfa aesar™
H66487MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 215.25
CAS 14108-60-2
MDL Number MFCD00070216
Chemical Name or Material 3-(2-Naphthyl)-DL-alanine
Physical Form Powder
Assay 98%
Storage Note 1 2° to 8°C
Synonym DL-Ala(2-naphthyl)-OH
Quantity 1g
Color Off-white
Safety and Handling

2° to 8°C