Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(2-Naphthyl)acrylic acid, 98%

$79.58 - $1,159.08

Chemical Identifiers

Chemical Name or Material 3-(2-Naphthyl)acrylic acid
CAS 51557-26-7
Molecular Formula C13H10O2
Formula Weight 198.22
MDL Number MFCD00209666
Synonym Naphthalene-2-acrylic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1671503

 
alfa aesar™
A1671503
1g Each for $79.58

AAA1671506

 
alfa aesar™
A1671506
5g Each for $280.15

AAA1671514

 
alfa aesar™
A1671514
25g Each for $1,159.08
Description & Specifications

Specifications

Formula Weight 198.22
Chemical Name or Material 3-(2-Naphthyl)acrylic acid
Assay 98%
Molecular Formula C13H10O2
MDL Number MFCD00209666
CAS 51557-26-7
Synonym Naphthalene-2-acrylic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.