missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(2-Methylphenylsulfamoyl)benzeneboronic acid, 97%

Manufacturer:  Alfa Aesar™ H54280MD

 View more versions of this product

Catalog No. AAH54280MD



Specifications

Specifications

3-(2-Methylphenylsulfamoyl)benzeneboronic acid
1449142-47-5
MFCD20265225
UBCMWUGADBYWIT-UHFFFAOYSA-N
[3-[(2-methylphenyl)sulfamoyl]phenyl]boronic acid
73995734
97%
250mg
C13H14BNO4S
3-2-methylphenyl sulfamoyl phenylboronic acid
B(C1=CC(=CC=C1)S(=O)(=O)NC2=CC=CC=C2C)(O)O
291.128
291.13

Chemical Identifiers

1449142-47-5
291.128
UBCMWUGADBYWIT-UHFFFAOYSA-N
73995734
B(C1=CC(=CC=C1)S(=O)(=O)NC2=CC=CC=C2C)(O)O
C13H14BNO4S
MFCD20265225
3-2-methylphenyl sulfamoyl phenylboronic acid
[3-[(2-methylphenyl)sulfamoyl]phenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents