missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(2-Methylphenyl)-5-phenyl-1,2,4-oxadiazole, 97%

Manufacturer:  Alfa Aesar™ H54927MD

 View more versions of this product

Catalog No. AAH54927MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

3-(2-Methylphenyl)-5-phenyl-1,2,4-oxadiazole
1400191-58-3
MFCD20265377
UPWQKWQWFBYGFM-UHFFFAOYSA-N
3-(2-methylphenyl)-5-phenyl-1,2,4-oxadiazole
54366090
97%
250mg
C15H12N2O
3-2-methylphenyl-5-phenyl-1,2,4-oxadiazole
CC1=CC=CC=C1C2=NOC(=N2)C3=CC=CC=C3
236.274
236.27

Chemical Identifiers

1400191-58-3
236.274
UPWQKWQWFBYGFM-UHFFFAOYSA-N
54366090
CC1=CC=CC=C1C2=NOC(=N2)C3=CC=CC=C3
C15H12N2O
MFCD20265377
3-2-methylphenyl-5-phenyl-1,2,4-oxadiazole
3-(2-methylphenyl)-5-phenyl-1,2,4-oxadiazole
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents