Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(2-Methylphenoxy)-1,2-propanediol, 98%

$59.96 - $705.16

Chemical Identifiers

Chemical Name or Material 3-(2-Methylphenoxy)-1,2-propanediol
CAS 59-47-2
Molecular Formula C10H14O3
Formula Weight 182.22
MDL Number MFCD00004718
Merck Index 14,5850
Synonym Mephenesin; 3-(o-Tolyloxy)-1,2-propanediol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2192814

  SDS
alfa aesar™
B2192814
25g Each for $59.96

AAB2192822

  SDS
alfa aesar™
B2192822
100g Each for $192.86

AAB2192836

  SDS
alfa aesar™
B2192836
500g Each for $705.16
Description & Specifications

Specifications

Formula Weight 182.22
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD00004718
Boiling Point 153° to 154°C (4mmHg)
Solubility Information Soluble in alcohol
Synonym Mephenesin; 3-(o-Tolyloxy)-1,2-propanediol
Assay 98%
Merck Index 14,5850
Quantity 25g
Molecular Formula C10H14O3
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Chemical Name or Material 3-(2-Methylphenoxy)-1,2-propanediol
Melting Point 69° to 71°C
CAS 59-47-2
Safety and Handling

Danger

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301
Toxic if swallowed.