Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(2-Methyl-4-nitro-1H-imidazol-1-yl)propionitrile, 97%

$40.69 - $562.03

Chemical Identifiers

Chemical Name or Material 3-(2-Methyl-4-nitro-1H-imidazol-1-yl)propionitrile
CAS 89128-08-5
Molecular Formula C7H8N4O2
Formula Weight 180.17
MDL Number MFCD00134458
Synonym 1-(2-Cyanoethyl)-2-methyl-4-nitroimidazole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2467503

  SDS
alfa aesar™
B2467503
1g Each for $40.69

AAB2467506

  SDS
alfa aesar™
B2467506
5g Each for $139.51

AAB2467514

  SDS
alfa aesar™
B2467514
25g Each for $562.03
Description & Specifications

Specifications

Formula Weight 180.17
CAS 89128-08-5
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 113° to 116°C
MDL Number MFCD00134458
Quantity 1g
Chemical Name or Material 3-(2-Methyl-4-nitro-1H-imidazol-1-yl)propionitrile
Assay 97%
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C7H8N4O2
Synonym 1-(2-Cyanoethyl)-2-methyl-4-nitroimidazole
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.