Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(2-Methyl-3-furylthio)-4-heptanone, 97%

$103.35 - $256.15

Chemical Identifiers

Chemical Name or Material 3-(2-Methyl-3-furylthio)-4-heptanone
CAS 61295-41-8
Molecular Formula C12H18O2S
Formula Weight 226.34
MDL Number MFCD01723735
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2737803

  SDS
alfa aesar™
H2737803
1g Each for $256.15

AAH27378MD

  SDS
alfa aesar™
H27378MD
250mg Each for $103.35
Description & Specifications

Specifications

Formula Weight 226.34
Assay 97%
Health Hazard 1 Warning
Quantity 1g
Health Hazard 2 GHS H Statement
H302
Harmful if swallowed.
Health Hazard 3 GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 3-(2-Methyl-3-furylthio)-4-heptanone
Molecular Formula C12H18O2S
MDL Number MFCD01723735
CAS 61295-41-8
Safety and Handling

Warning

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302
Harmful if swallowed.