missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(2-Methyl-1H-imidazol-1-yl)benzoic acid, 90%, Maybridge

Chemical Identifiers

CAS 898289-59-3
CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Molecular Formula C11H10N2O2
Formula Weight 202.21
MDL Number MFCD09065014
Chemical Name or Material 3-(2-Methyl-1H-imidazol-1-yl)benzoic acid
Assay 90%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC61301CB SDS Fisher Scientific Company
CC61301CB
Amber Glass Bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC61301DA SDS Fisher Scientific Company
CC61301DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

898289-59-3
100.0
C11H10N2O2
MFCD09065014
90%
89.5
90%
202.21
3-(2-Methyl-1H-imidazol-1-yl)benzoic acid

Specifications

202.21
100.0
90%
Amber Glass Bottle
898289-59-3
90%
250mg
MFCD09065014
89.5
C11H10N2O2
3-(2-Methyl-1H-imidazol-1-yl)benzoic acid

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.