missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-[(2-Methyl-1-piperidinyl)sulfonyl]benzeneboronic acid, 97%, Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific H5412703

 View more versions of this product

Catalog No. AAH5412703



Specifications

Specifications

3-[(2-Methyl-1-piperidinyl)sulfonyl]benzeneboronic acid
1449145-16-7
MFCD20265185
IZHIWXZQLZLQGQ-UHFFFAOYSA-N
[3-(2-methylpiperidin-1-yl)sulfonylphenyl]boronic acid
73995648
97%
1g
C12H18BNO4S
3-2-methyl-1-piperidinyl sulfonyl benzeneboronic acid
B(C1=CC(=CC=C1)S(=O)(=O)N2CCCCC2C)(O)O
283.149
283.15

Chemical Identifiers

1449145-16-7
283.149
IZHIWXZQLZLQGQ-UHFFFAOYSA-N
73995648
B(C1=CC(=CC=C1)S(=O)(=O)N2CCCCC2C)(O)O
C12H18BNO4S
MFCD20265185
3-2-methyl-1-piperidinyl sulfonyl benzeneboronic acid
[3-(2-methylpiperidin-1-yl)sulfonylphenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents