missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(2-Methyl-1,3-dioxolan-2-yl)benzeneboronic acid, 97%

Manufacturer:  Alfa Aesar™ H5328603

 View more versions of this product

Catalog No. AAH5328603



Specifications

Specifications

3-(2-Methyl-1,3-dioxolan-2-yl)benzeneboronic acid
1g
C10H13BO4
3-2-methyl-1,3-dioxolan-2-yl phenylboronic acid, 3-2-methyl-1,3-dioxolan-2-yl phenyl boronic acid, 2-3-boronobenzene-2-methyl-1,3-dioxolane, 2-3-boronophenyl-2-methyl-1,3-dioxolane, 3-2-methyl-1,3-dioxolan-2-yl benzeneboronic acid, 3-2-methyl-1,3-dioxolan-2-yl phenyl boronic acid, 3-boronoacetophenone ethylene acetal, boronic acid,b-3-2-methyl-1,3-dioxolan-2-yl phenyl
B(C1=CC(=CC=C1)C2(OCCO2)C)(O)O
208.02
208.02
100°C to 104°C
850568-50-2
MFCD04973736
NSDRSKQWUQJUAE-UHFFFAOYSA-N
[3-(2-methyl-1,3-dioxolan-2-yl)phenyl]boronic acid
3674880
97%

Chemical Identifiers

850568-50-2
208.02
NSDRSKQWUQJUAE-UHFFFAOYSA-N
3674880
B(C1=CC(=CC=C1)C2(OCCO2)C)(O)O
C10H13BO4
MFCD04973736
3-2-methyl-1,3-dioxolan-2-yl phenylboronic acid, 3-2-methyl-1,3-dioxolan-2-yl phenyl boronic acid, 2-3-boronobenzene-2-methyl-1,3-dioxolane, 2-3-boronophenyl-2-methyl-1,3-dioxolane, 3-2-methyl-1,3-dioxolan-2-yl benzeneboronic acid, 3-2-methyl-1,3-dioxolan-2-yl phenyl boronic acid, 3-boronoacetophenone ethylene acetal, boronic acid,b-3-2-methyl-1,3-dioxolan-2-yl phenyl
[3-(2-methyl-1,3-dioxolan-2-yl)phenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents