missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(2-Methoxyphenoxy)piperidine

$85.74 - $306.31

Chemical Identifiers

CAS 902837-27-8
Molecular Formula C12H17NO2
Formula Weight 207.27
MDL Number MFCD08061100
Chemical Name or Material 3-(2-Methoxyphenoxy)piperidine
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5098803 SDS Alfa Aesar™
H5098803
1g Each for $306.31
AAH50988MD SDS Alfa Aesar™
H50988MD
250mg Each for $85.74
Specifications Description & Specifications

Chemical Identifiers

902837-27-8
207.27
3-(2-Methoxyphenoxy)piperidine
C12H17NO2
MFCD08061100

Specifications

902837-27-8
207.27
1g
C12H17NO2
MFCD08061100
3-(2-Methoxyphenoxy)piperidine

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning