Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(2-Methoxyethylaminocarbonyl)benzeneboronic acid pinacol ester, 97%

$116.03 - $355.59

Chemical Identifiers

Chemical Name or Material 3-(2-Methoxyethylaminocarbonyl)benzeneboronic acid pinacol ester
Molecular Formula C16H24BNO4
Formula Weight 305.18
MDL Number MFCD09266187
Synonym 3-(2-Methoxyethylaminocarbonyl)phenylboronic acid pinacol ester; N-(2-Methoxyethyl)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2834003

  SDS
alfa aesar™
H2834003
1g Each for $116.03

AAH2834006

  SDS
alfa aesar™
H2834006
5g Each for $355.59
Description & Specifications

Specifications

Formula Weight 305.18
Assay 97%
Melting Point 93° to 98°C
Quantity 1g
MDL Number MFCD09266187
Chemical Name or Material 3-(2-Methoxyethylaminocarbonyl)benzeneboronic acid pinacol ester
Synonym 3-(2-Methoxyethylaminocarbonyl)phenylboronic acid pinacol ester; N-(2-Methoxyethyl)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
Molecular Formula C16H24BNO4