missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(2-Methoxyethoxy)propyl Bromide, TCI America™

$201.67 - $660.02

Chemical Identifiers

CAS 59551-75-6
Molecular Formula C6H13BrO2
Molecular Weight (g/mol) 197.072
MDL Number MFCD01321130
InChI Key XXRSNYDVVFLAPC-UHFFFAOYSA-N
Synonym 1-Bromo-3-(2-methoxyethoxy)propane, Ethylene Glycol 3-Bromopropyl Methyl Ether
PubChem CID 15636133
IUPAC Name 1-bromo-3-(2-methoxyethoxy)propane
SMILES COCCOCCCBr
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B18421G View Documents TCI America
B18421G
1g Each for $201.67
 
B18425G View Documents TCI America
B18425G
5g Each for $660.02
 
Specifications

Chemical Identifiers

59551-75-6
197.072
XXRSNYDVVFLAPC-UHFFFAOYSA-N
15636133
COCCOCCCBr
C6H13BrO2
MFCD01321130
1-Bromo-3-(2-methoxyethoxy)propane, Ethylene Glycol 3-Bromopropyl Methyl Ether
1-bromo-3-(2-methoxyethoxy)propane

Specifications

Yellow
59551-75-6
MFCD01321130
1-Bromo-3-(2-methoxyethoxy)propane, Ethylene Glycol 3-Bromopropyl Methyl Ether
COCCOCCCBr
197.072
197.07
≥95.0% (GC)
1g
C6H13BrO2
1993
XXRSNYDVVFLAPC-UHFFFAOYSA-N
1-bromo-3-(2-methoxyethoxy)propane
15636133
Clear Liquid at 20°C
3-(2-Methoxyethoxy)propyl Bromide
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator