missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(2-Isopropylphenoxy)piperidine

$80.96 - $80.96

Chemical Identifiers

CAS 902837-35-8
Molecular Formula C14H21NO
Formula Weight 219.33
MDL Number MFCD08061106
Chemical Name or Material 3-(2-Isopropylphenoxy)piperidine
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH50996MD SDS Alfa Aesar™
H50996MD
250mg Each for $80.96
AAH5099603 SDS Alfa Aesar™
H5099603
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

902837-35-8
219.33
3-(2-Isopropylphenoxy)piperidine
C14H21NO
MFCD08061106

Specifications

250mg
C14H21NO
MFCD08061106
902837-35-8
219.33
3-(2-Isopropylphenoxy)piperidine

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning