missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-(2-Hydroxyethoxy)benzaldehyde 96.0+%, TCI America™
$94.56 - $283.68
Chemical Identifiers
CAS | 60345-97-3 |
---|---|
Molecular Formula | C9H10O3 |
Molecular Weight (g/mol) | 166.18 |
MDL Number | MFCD00191448 |
InChI Key | LIUCHRXQRHVSJI-UHFFFAOYSA-N |
PubChem CID | 6453965 |
IUPAC Name | 3-(2-hydroxyethoxy)benzaldehyde |
SMILES | OCCOC1=CC=CC(C=O)=C1 |
Chemical Identifiers
60345-97-3 | |
166.18 | |
LIUCHRXQRHVSJI-UHFFFAOYSA-N | |
3-(2-hydroxyethoxy)benzaldehyde |
C9H10O3 | |
MFCD00191448 | |
6453965 | |
OCCOC1=CC=CC(C=O)=C1 |
Specifications
60345-97-3 | |
C9H10O3 | |
5 g | |
OCCOC1=CC=CC(C=O)=C1 | |
166.18 | |
166.18 | |
Liquid |
Yellow | |
MFCD00191448 | |
LIUCHRXQRHVSJI-UHFFFAOYSA-N | |
3-(2-hydroxyethoxy)benzaldehyde | |
6453965 | |
≥96.0% (GC) | |
3-(2-Hydroxyethoxy)benzaldehyde |
Safety and Handling
TSCA : Yes
Recommended Storage : Refrigerator