missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(2-Hydroxy-2-propanyl)phenylboronic acid pinacol ester, 97%, ACROS Organics™

Chemical Identifiers

CAS 1309980-11-7
Molecular Formula C15H23BO3
Molecular Weight (g/mol) 262.156
InChI Key NNPQWBXXZHNLAW-UHFFFAOYSA-N
Synonym 2-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl propan-2-ol, 3-2-hydroxypropan-2-yl phenylboronic acid pinacol ester, 2-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl propan-2-ol, 3-2-hydroxy-2-propyl phenylboronic acid pinacol ester, 3-2-hydroxy-2-propanyl phenylboronic acid pinacol ester, 3-2-hydroxy-2-propanyl phenylboronic acid pinacol ester, 3-2-hydroxypropan-2-yl phenyl boronic acid pinacol ester, 2-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl phenyl propan-2-ol, 2-3-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl propan-2-ol
PubChem CID 46738028
IUPAC Name 2-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]propan-2-ol
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)C(C)(C)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC445070010 SDS View Documents Acros Organics
AC445070010
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

1309980-11-7
262.156
2-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl propan-2-ol, 3-2-hydroxypropan-2-yl phenylboronic acid pinacol ester, 2-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl propan-2-ol, 3-2-hydroxy-2-propyl phenylboronic acid pinacol ester, 3-2-hydroxy-2-propanyl phenylboronic acid pinacol ester, 3-2-hydroxy-2-propanyl phenylboronic acid pinacol ester, 3-2-hydroxypropan-2-yl phenyl boronic acid pinacol ester, 2-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl phenyl propan-2-ol, 2-3-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl propan-2-ol
2-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]propan-2-ol
C15H23BO3
NNPQWBXXZHNLAW-UHFFFAOYSA-N
46738028
B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)C(C)(C)O

Specifications

Glass bottle
1g
97%
2-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl propan-2-ol, 3-2-hydroxypropan-2-yl phenylboronic acid pinacol ester, 2-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl propan-2-ol, 3-2-hydroxy-2-propyl phenylboronic acid pinacol ester, 3-2-hydroxy-2-propanyl phenylboronic acid pinacol ester, 3-2-hydroxy-2-propanyl phenylboronic acid pinacol ester, 3-2-hydroxypropan-2-yl phenyl boronic acid pinacol ester, 2-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl phenyl propan-2-ol, 2-3-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl propan-2-ol
NNPQWBXXZHNLAW-UHFFFAOYSA-N
2-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]propan-2-ol
46738028
97%
69.0°C to 70.0°C
1309980-11-7
C15H23BO3
Solubility in water: insoluble.
B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)C(C)(C)O
262.156
262.16
3-(2-Hydroxy-2-propanyl)phenylboronic acid pinacol ester