missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(2-Hydroxy-2-propanyl)phenylboronic acid pinacol ester, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC445070010

 View more versions of this product

Catalog No. AC445070010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

3-(2-Hydroxy-2-propanyl)phenylboronic acid pinacol ester
69.0°C to 70.0°C
1309980-11-7
C15H23BO3
Solubility in water: insoluble.
B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)C(C)(C)O
262.16
262.16
Glass bottle
1g
96% min. (GC)
2-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl phenyl propan-2-ol, 2-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl propan-2-ol, 2-3-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl propan-2-ol, 3-2-hydroxy-2-propanyl phenylboronic acid pinacol ester, 3-2-hydroxy-2-propyl phenylboronic acid pinacol ester, 3-2-hydroxypropan-2-yl phenyl boronic acid pinacol ester, 3-2-hydroxypropan-2-yl phenylboronic acid pinacol ester
NNPQWBXXZHNLAW-UHFFFAOYSA-N
2-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]propan-2-ol
46738028
97%

Chemical Identifiers

1309980-11-7
262.16
2-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl phenyl propan-2-ol, 2-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl propan-2-ol, 2-3-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl propan-2-ol, 3-2-hydroxy-2-propanyl phenylboronic acid pinacol ester, 3-2-hydroxy-2-propyl phenylboronic acid pinacol ester, 3-2-hydroxypropan-2-yl phenyl boronic acid pinacol ester, 3-2-hydroxypropan-2-yl phenylboronic acid pinacol ester
2-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]propan-2-ol
C15H23BO3
NNPQWBXXZHNLAW-UHFFFAOYSA-N
46738028
B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)C(C)(C)O
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER

GHS Signal Word: Warning

SDS Documents