Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(2-Furyl)propionic acid, 97%

$150.68 - $150.68

Chemical Identifiers

Chemical Name or Material 3-(2-Furyl)propionic acid
CAS 935-13-7
Molecular Formula C7H8O3
Formula Weight 140.14
MDL Number MFCD00005346
Synonym 2-Furanpropionic acid; 2-Furanpropionic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2727306

  SDS
alfa aesar™
H2727306
5g Each for $150.68
Description & Specifications

Specifications

Formula Weight 140.14
Quantity 5g
Synonym 2-Furanpropionic acid; 2-Furanpropionic acid
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 56° to 60°C
Chemical Name or Material 3-(2-Furyl)propionic acid
MDL Number MFCD00005346
CAS 935-13-7
Molecular Formula C7H8O3
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
Assay 97%
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.