missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(2-Furyl)acrylic Acid, TCI America™

Manufacturer:  TCI America F008410G

 View more versions of this product

Catalog No. F008410G



Specifications

Specifications

3-(2-Furyl)acrylic Acid
Yellow
10g
C7H6O3
3-2-furyl acrylic acid, e-3-furan-2-yl acrylic acid, 2-furanacrylic acid, 3-furan-2-yl acrylic acid, furanacrylic acid, 2-furalacetic acid, furacrylic acid, furylacrylic acid, 2e-3-furan-2-yl prop-2-enoic acid, 2-propenoic acid, 3-2-furanyl
C1=COC(=C1)C=CC(=O)O
138.122
138.12
≥98.0% (GC,T)
286°C
141°C
539-47-9
MFCD00003257
ZCJLOOJRNPHKAV-ONEGZZNKSA-N
(E)-3-(furan-2-yl)prop-2-enoic acid
643402
Crystal-Powder at 20°C

Chemical Identifiers

539-47-9
138.122
ZCJLOOJRNPHKAV-ONEGZZNKSA-N
643402
C1=COC(=C1)C=CC(=O)O
C7H6O3
MFCD00003257
3-2-furyl acrylic acid, e-3-furan-2-yl acrylic acid, 2-furanacrylic acid, 3-furan-2-yl acrylic acid, furanacrylic acid, 2-furalacetic acid, furacrylic acid, furylacrylic acid, 2e-3-furan-2-yl prop-2-enoic acid, 2-propenoic acid, 3-2-furanyl
(E)-3-(furan-2-yl)prop-2-enoic acid
Safety and Handling

Safety and Handling

EINECSNumber : (9)-1158

RTECSNumber : LT8560000

TSCA : Yes

Documents