Please login to your online account to display your discounted pricing

[3-(2-Furyl)-1-methyl-1H-pyrazol-5-yl]methanol 97+%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC47009CB

 View more versions of this product

Catalog No. CC47009CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 178.19
Quantity 250mg
Assay 97%
Chemical Name or Material [3-(2-Furyl)-1-methyl-1H-pyrazol-5-yl]methanol
Molecular Formula C9H10N2O2
CAS 886851-33-8
Assay Percent Range 97%
Packaging Amber Glass Bottle

Specifications

Chemical Name or Material [3-(2-Furyl)-1-methyl-1H-pyrazol-5-yl]methanol
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H10N2O2
Formula Weight 178.19