Please login to your online account to display your discounted pricing

3-(2-Formyl-1H-pyrrol-1-yl)propanenitrile, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 3-(2-Formyl-1H-pyrrol-1-yl)propanenitrile
CAS 43036-05-1
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C8H8N2O
Formula Weight 148.16
MDL Number MFCD02180764
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SP01383EA

  SDS
fisher scientific company
SP01383EA
Amber Glass Bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 148.16
Quantity 10g
Assay 97%
CAS Min % 97.0
Molecular Formula C8H8N2O
Chemical Name or Material 3-(2-Formyl-1H-pyrrol-1-yl)propanenitrile
CAS 43036-05-1
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD02180764
Packaging Amber Glass Bottle
Assay Percent Range 97%
Health Hazard 1 GHS Signal Word: Warning