missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(2-Fluorophenylsulfamoyl)benzeneboronic acid, 97%

$315.01 - $1,004.50

Chemical Identifiers

CAS 1449142-51-1
Molecular Formula C12H11BFNO4S
Molecular Weight (g/mol) 295.091
MDL Number MFCD20265270
InChI Key CSHNEUKEFGIIOZ-UHFFFAOYSA-N
Synonym 3-2-fluorophenyl sulfamoyl phenylboronic acid
PubChem CID 73995813
IUPAC Name [3-[(2-fluorophenyl)sulfamoyl]phenyl]boronic acid
SMILES B(C1=CC(=CC=C1)S(=O)(=O)NC2=CC=CC=C2F)(O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH54450MD View Documents Alfa Aesar™
H54450MD
250mg Each for $315.01
 
AAH5445003 View Documents Alfa Aesar™
H5445003
1g Each for $1,004.50
 
Specifications

Chemical Identifiers

1449142-51-1
295.091
CSHNEUKEFGIIOZ-UHFFFAOYSA-N
73995813
B(C1=CC(=CC=C1)S(=O)(=O)NC2=CC=CC=C2F)(O)O
C12H11BFNO4S
MFCD20265270
3-2-fluorophenyl sulfamoyl phenylboronic acid
[3-[(2-fluorophenyl)sulfamoyl]phenyl]boronic acid

Specifications

250mg
C12H11BFNO4S
3-2-fluorophenyl sulfamoyl phenylboronic acid
B(C1=CC(=CC=C1)S(=O)(=O)NC2=CC=CC=C2F)(O)O
295.091
295.09
3-(2-Fluorophenylsulfamoyl)benzeneboronic acid
1449142-51-1
MFCD20265270
CSHNEUKEFGIIOZ-UHFFFAOYSA-N
[3-[(2-fluorophenyl)sulfamoyl]phenyl]boronic acid
73995813
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures