missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(2-Fluorophenylsulfamoyl)benzeneboronic acid, 97%

Manufacturer:  Alfa Aesar™ H5445003

 View more versions of this product

Catalog No. AAH5445003



Specifications

Specifications

3-(2-Fluorophenylsulfamoyl)benzeneboronic acid
1449142-51-1
MFCD20265270
CSHNEUKEFGIIOZ-UHFFFAOYSA-N
[3-[(2-fluorophenyl)sulfamoyl]phenyl]boronic acid
73995813
97%
1g
C12H11BFNO4S
3-2-fluorophenyl sulfamoyl phenylboronic acid
B(C1=CC(=CC=C1)S(=O)(=O)NC2=CC=CC=C2F)(O)O
295.091
295.09

Chemical Identifiers

1449142-51-1
295.091
CSHNEUKEFGIIOZ-UHFFFAOYSA-N
73995813
B(C1=CC(=CC=C1)S(=O)(=O)NC2=CC=CC=C2F)(O)O
C12H11BFNO4S
MFCD20265270
3-2-fluorophenyl sulfamoyl phenylboronic acid
[3-[(2-fluorophenyl)sulfamoyl]phenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents