missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-[2-(Diethylamino)ethylcarbamoyl]benzeneboronic acid hydrochloride, 97%, Alfa Aesar™

Manufacturer:  Alfa Aesar H5328103

 View more versions of this product

Catalog No. AAH5328103



Specifications

Specifications

3-[2-(Diethylamino)ethylcarbamoyl]benzeneboronic acid hydrochloride
957061-01-7
MFCD09027256
CXICYVWYLASJEJ-UHFFFAOYSA-N
[3-[2-(diethylamino)ethylcarbamoyl]phenyl]boronic acid;hydrochloride
44119693
97%
1g
C13H22BClN2O3
3-2-diethylamino ethyl carbamoyl phenyl boronic acid hydrochloride, 3-2-n,n-diethylaminoethylaminocarbonyl phenylboronic acid, hcl, 3-2-diethylamino ethyl carbamoyl benzeneboronic acid hydrochloride, 3-2-diethylamino ethyl carbamoyl phenylboronic acid hydrochloride, 3-2-n,n-diethylaminoethylaminocarbonyl phenylboronic acid hcl, 3-2-diethylamino ethyl carbamoyl phenyl boronicacidhydrochloride, 3-2-diethylamino ethyl carbamoyl-phenyl boronic acid hydrochloride, 3-2-diethylamino ethyl carbamoyl phenyl boronic acid-hydrogen chloride 1/1
B(C1=CC(=CC=C1)C(=O)NCCN(CC)CC)(O)O.Cl
300.59
300.59

Chemical Identifiers

957061-01-7
300.59
CXICYVWYLASJEJ-UHFFFAOYSA-N
44119693
B(C1=CC(=CC=C1)C(=O)NCCN(CC)CC)(O)O.Cl
C13H22BClN2O3
MFCD09027256
3-2-diethylamino ethyl carbamoyl phenyl boronic acid hydrochloride, 3-2-n,n-diethylaminoethylaminocarbonyl phenylboronic acid, hcl, 3-2-diethylamino ethyl carbamoyl benzeneboronic acid hydrochloride, 3-2-diethylamino ethyl carbamoyl phenylboronic acid hydrochloride, 3-2-n,n-diethylaminoethylaminocarbonyl phenylboronic acid hcl, 3-2-diethylamino ethyl carbamoyl phenyl boronicacidhydrochloride, 3-2-diethylamino ethyl carbamoyl-phenyl boronic acid hydrochloride, 3-2-diethylamino ethyl carbamoyl phenyl boronic acid-hydrogen chloride 1/1
[3-[2-(diethylamino)ethylcarbamoyl]phenyl]boronic acid;hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents