Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(2-Chlorophenyl)-5-methylisoxazole-4-carbonyl chloride, 98+%

Chemical Identifiers

Chemical Name or Material 3-(2-Chlorophenyl)-5-methylisoxazole-4-carbonyl chloride
CAS 25629-50-9
Molecular Formula C11H7Cl2NO2
Formula Weight 256.09
MDL Number MFCD00020811
UN Number UN3261
Beilstein 791931
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0987006

  SDS
alfa aesar™
L0987006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0987018

  SDS
alfa aesar™
L0987018
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0987030

  SDS
alfa aesar™
L0987030
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 256.09
Chemical Name or Material 3-(2-Chlorophenyl)-5-methylisoxazole-4-carbonyl chloride
CAS 25629-50-9
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Assay ≥98%
Health Hazard 1 Danger
MDL Number MFCD00020811
Quantity 5g
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA TSCA
Beilstein 791931
UN Number UN3261
Melting Point 41° to 42°C
Molecular Formula C11H7Cl2NO2
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.