Please login to your online account to display your discounted pricing

3-(2-chloro-6-fluorophenyl)-5-methylisoxazole-4-carboxamide, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  RF00732DA

 View more versions of this product

Catalog No. RF00732DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 254.65
Quantity 1g
Assay 97%
CAS 918896
CAS Min % 97.0
Chemical Name or Material 3-(2-Chloro-6-fluorophenyl)-5-methylisoxazole-4-carboxamide
Molecular Formula C11H8ClFN2O2
Packaging Amber Glass Bottle
Assay Percent Range 97%
MDL Number MFCD00173954

Specifications

Chemical Name or Material 3-(2-Chloro-6-fluorophenyl)-5-methylisoxazole-4-carboxamide
CAS 918896
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C11H8ClFN2O2
Formula Weight 254.65
MDL Number MFCD00173954