Please login to your online account to display your discounted pricing

3-(2-chloro-6-fluorophenyl)-5-methylisoxazole-4-carboxamide, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 3-(2-Chloro-6-fluorophenyl)-5-methylisoxazole-4-carboxamide
CAS 918896
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C11H8ClFN2O2
Formula Weight 254.65
MDL Number MFCD00173954
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

RF00732DA

  SDS
fisher scientific company
RF00732DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

RF00732EA

  SDS
fisher scientific company
RF00732EA
Amber Glass Bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 254.65
Quantity 1g
Assay 97%
CAS 918896
CAS Min % 97.0
Chemical Name or Material 3-(2-Chloro-6-fluorophenyl)-5-methylisoxazole-4-carboxamide
Molecular Formula C11H8ClFN2O2
Packaging Amber Glass Bottle
Assay Percent Range 97%
MDL Number MFCD00173954