Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(2-Chloro-5-methylphenoxy)piperidine, 98%

$80.96 - $292.46

Chemical Identifiers

Chemical Name or Material 3-(2-Chloro-5-methylphenoxy)piperidine
CAS 902837-30-3
Molecular Formula C12H16ClNO
Formula Weight 225.72
MDL Number MFCD08061103
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5099103

 
alfa aesar™
H5099103
1g Each for $292.46

AAH50991MD

 
alfa aesar™
H50991MD
250mg Each for $80.96
Description & Specifications

Specifications

Formula Weight 225.72
Chemical Name or Material 3-(2-Chloro-5-methylphenoxy)piperidine
Assay 98%
Molecular Formula C12H16ClNO
MDL Number MFCD08061103
CAS 902837-30-3
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.