missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(2-Carboxyvinyl)benzeneboronic acid, 97%

Manufacturer:  Alfa Aesar™ L16369MD

 View more versions of this product

Catalog No. AAL16369MD



Specifications

Specifications

3-(2-Carboxyvinyl)benzeneboronic acid
250mg
C9H9BO4
2e-3-3-dihydroxyboranyl phenyl prop-2-enoic acid, 3--e-2-carboxyvinyl benzeneboronic acid, 3-2-carboxyvinyl benzeneboronic acid, 3-2-carboxyvinyl phenylboronic acid, 3-3-boronophenyl acrylic acid, 3-3-dihydroxyboranyl phenyl prop-2-enoic acid, 3-boronocinnamic acid, 3-phenylpropylester boronic acid, 3-trans-2-carboxyvinyl phenylboronic acid, e-3-3-boronophenyl acrylic acid
B(C1=CC(=CC=C1)C=CC(=O)O)(O)O
191.977
191.98
~218°C
216144-91-1
MFCD01075738
QCHIEOGZUMAQKI-SNAWJCMRSA-N
(E)-3-(3-boronophenyl)prop-2-enoic acid
5844876
97%

Chemical Identifiers

216144-91-1
191.977
QCHIEOGZUMAQKI-SNAWJCMRSA-N
5844876
B(C1=CC(=CC=C1)C=CC(=O)O)(O)O
C9H9BO4
MFCD01075738
2e-3-3-dihydroxyboranyl phenyl prop-2-enoic acid, 3--e-2-carboxyvinyl benzeneboronic acid, 3-2-carboxyvinyl benzeneboronic acid, 3-2-carboxyvinyl phenylboronic acid, 3-3-boronophenyl acrylic acid, 3-3-dihydroxyboranyl phenyl prop-2-enoic acid, 3-boronocinnamic acid, 3-phenylpropylester boronic acid, 3-trans-2-carboxyvinyl phenylboronic acid, e-3-3-boronophenyl acrylic acid
(E)-3-(3-boronophenyl)prop-2-enoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

Documents