missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(2-Carboxyvinyl)benzeneboronic acid, 97%

Manufacturer:  Alfa Aesar™ L1636903

 View more versions of this product

Catalog No. AAL1636903



Specifications

Specifications

3-(2-Carboxyvinyl)benzeneboronic acid
1g
C9H9BO4
2e-3-3-dihydroxyboranyl phenyl prop-2-enoic acid
B(C1=CC(=CC=C1)C=CC(=O)O)(O)O
191.977
191.98
~218°C
216144-91-1
MFCD01075738
QCHIEOGZUMAQKI-SNAWJCMRSA-N
(E)-3-(3-boronophenyl)prop-2-enoic acid
5844876
97%

Chemical Identifiers

216144-91-1
191.977
QCHIEOGZUMAQKI-SNAWJCMRSA-N
5844876
B(C1=CC(=CC=C1)C=CC(=O)O)(O)O
C9H9BO4
MFCD01075738
2e-3-3-dihydroxyboranyl phenyl prop-2-enoic acid
(E)-3-(3-boronophenyl)prop-2-enoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents