Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(2-Bromophenyl)propionic acid, 96%

$60.34 - $60.34

Chemical Identifiers

Chemical Name or Material 3-(2-Bromophenyl)propionic acid
CAS 15115-58-9
Molecular Formula C9H9BrO2
Formula Weight 229.07
MDL Number MFCD01310791
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3393306

 
alfa aesar™
H3393306*
5g Each for $60.34
Description & Specifications

Specifications

Formula Weight 229.07
Chemical Name or Material 3-(2-Bromophenyl)propionic acid
Assay 96%
Molecular Formula C9H9BrO2
MDL Number MFCD01310791
CAS 15115-58-9
Melting Point 93° to 95°C
Quantity 5g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.