missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(2-Bromophenyl)-1-propanol, 98%

$71.08 - $845.08

Chemical Identifiers

CAS 52221-92-8
Molecular Formula C9H11BrO
Molecular Weight (g/mol) 215.09
MDL Number MFCD09028685
InChI Key VODAJGPTULSNSU-UHFFFAOYSA-N
Synonym 3-2-bromo-phenyl-propan-1-ol, 3-2-bromophenyl propan-1-ol, 3-2-bromophenyl-1-propanol, benzenepropanol, 2-bromo, ksc268e5r, 3-2-bromo-phenyl ;-propan-1-ol
PubChem CID 10512846
IUPAC Name 3-(2-bromophenyl)propan-1-ol
SMILES C1=CC=C(C(=C1)CCCO)Br
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH63884MD View Documents Alfa Aesar™
H63884MD
250mg Each for $71.08
 
AAH6388403 View Documents Alfa Aesar™
H6388403
1g Each for $211.27
 
AAH6388406 View Documents Alfa Aesar™
H6388406
5g Each for $845.08
 
Specifications

Chemical Identifiers

52221-92-8
215.09
VODAJGPTULSNSU-UHFFFAOYSA-N
10512846
C1=CC=C(C(=C1)CCCO)Br
C9H11BrO
MFCD09028685
3-2-bromo-phenyl-propan-1-ol, 3-2-bromophenyl propan-1-ol, 3-2-bromophenyl-1-propanol, benzenepropanol, 2-bromo, ksc268e5r, 3-2-bromo-phenyl ;-propan-1-ol
3-(2-bromophenyl)propan-1-ol

Specifications

Colorless
52221-92-8
MFCD09028685
VODAJGPTULSNSU-UHFFFAOYSA-N
3-(2-bromophenyl)propan-1-ol
10512846
Liquid
3-(2-Bromophenyl)-1-propanol
250mg
C9H11BrO
3-2-bromo-phenyl-propan-1-ol, 3-2-bromophenyl propan-1-ol, 3-2-bromophenyl-1-propanol, benzenepropanol, 2-bromo, ksc268e5r, 3-2-bromo-phenyl ;-propan-1-ol
C1=CC=C(C(=C1)CCCO)Br
215.09
215.09
98%