missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(2-Bromophenyl)-1-propanol, 98%

Manufacturer:  Alfa Aesar™ H6388406

 View more versions of this product

Catalog No. AAH6388406



Specifications

Specifications

3-(2-Bromophenyl)-1-propanol
5g
C9H11BrO
3-2-bromo-phenyl-propan-1-ol, 3-2-bromophenyl propan-1-ol, 3-2-bromophenyl-1-propanol, benzenepropanol, 2-bromo, ksc268e5r, 3-2-bromo-phenyl ;-propan-1-ol
C1=CC=C(C(=C1)CCCO)Br
215.09
215.09
98%
Colorless
52221-92-8
MFCD09028685
VODAJGPTULSNSU-UHFFFAOYSA-N
3-(2-bromophenyl)propan-1-ol
10512846
Liquid

Chemical Identifiers

52221-92-8
215.09
VODAJGPTULSNSU-UHFFFAOYSA-N
10512846
C1=CC=C(C(=C1)CCCO)Br
C9H11BrO
MFCD09028685
3-2-bromo-phenyl-propan-1-ol, 3-2-bromophenyl propan-1-ol, 3-2-bromophenyl-1-propanol, benzenepropanol, 2-bromo, ksc268e5r, 3-2-bromo-phenyl ;-propan-1-ol
3-(2-bromophenyl)propan-1-ol
Documents