missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(2-Bromophenoxy)-N,N-dimethylpropylamine, 97%, Maybridge

Manufacturer:  Fisher Scientific Company CC59610DE

 View more versions of this product

Catalog No. CC59610DE

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

258.16
97%
3-(2-Bromophenoxy)-N, N-dimethylpropylamine
97%
Amber Glass Bottle
C11H16BrNO
5g
100.0
MFCD03718246
96.5
915707-52-7

Chemical Identifiers

915707-52-7
100.0
C11H16BrNO
MFCD03718246
97%
96.5
97%
258.16
3-(2-Bromophenoxy)-N, N-dimethylpropylamine

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger