missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-(2-Bromoethyl)indole, 95%, Thermo Scientific™
Chemical Identifiers
CAS | 3389-21-7 |
---|---|
Molecular Formula | C10H10BrN |
Molecular Weight (g/mol) | 224.1 |
MDL Number | MFCD00130155 |
InChI Key | NTLAICDKHHQUGC-UHFFFAOYSA-N |
Synonym | 3-2-bromoethyl-1h-indole, 3-2-bromoethyl indole, 1h-indole, 3-2-bromoethyl, 3-2-bromo-ethyl-1h-indole, indol-3-ylethylbromide, 3-2-bromethyl-indol, acmc-1c2el, 3-2-bromoethyl-indole, 3-2-bromo-ethyl-indole, chembl87532 |
PubChem CID | 76924 |
IUPAC Name | 3-(2-bromoethyl)-1H-indole |
SMILES | C1=CC=C2C(=C1)C(=CN2)CCBr |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC426910010
|
Thermo Scientific Chemicals
426910010 |
1g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC426910050
|
Thermo Scientific Chemicals
426910050 |
5g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
3389-21-7 | |
224.1 | |
NTLAICDKHHQUGC-UHFFFAOYSA-N | |
76924 | |
C1=CC=C2C(=C1)C(=CN2)CCBr |
C10H10BrN | |
MFCD00130155 | |
3-2-bromoethyl-1h-indole, 3-2-bromoethyl indole, 1h-indole, 3-2-bromoethyl, 3-2-bromo-ethyl-1h-indole, indol-3-ylethylbromide, 3-2-bromethyl-indol, acmc-1c2el, 3-2-bromoethyl-indole, 3-2-bromo-ethyl-indole, chembl87532 | |
3-(2-bromoethyl)-1H-indole |
Specifications
3389-21-7 | |
100.0 | |
94% min. (GC) | |
C10H10BrN | |
1g | |
NTLAICDKHHQUGC-UHFFFAOYSA-N | |
3-(2-bromoethyl)-1H-indole | |
76924 | |
95% |
96.0 | |
Authentic | |
Glass bottle | |
MFCD00130155 | |
3-2-bromoethyl-1h-indole, 3-2-bromoethyl indole, 1h-indole, 3-2-bromoethyl, 3-2-bromo-ethyl-1h-indole, indol-3-ylethylbromide, 3-2-bromethyl-indol, acmc-1c2el, 3-2-bromoethyl-indole, 3-2-bromo-ethyl-indole, chembl87532 | |
C1=CC=C2C(=C1)C(=CN2)CCBr | |
224.1 | |
224.1 | |
3-(2-Bromoethyl)indole |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with
GHS Signal Word: Danger
EINECSNumber : 222-219-2
RUO â Research Use Only