missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(2-bromo-5-methoxyphenyl)-3-oxopropanenitrile, Maybridge

Chemical Identifiers

Assay Percent Range 97%
Molecular Formula C10H8BrNO2
Formula Weight 254.08
Chemical Name or Material 3-(2-Bromo-5-methoxyphenyl)-3-oxopropanenitrile
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CD05258EB SDS Fisher Scientific Company
CD05258EB
Amber glass bottle 25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CD05258DA SDS Fisher Scientific Company
CD05258DA
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CD05258EA SDS Fisher Scientific Company
CD05258EA
10g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

97%
254.08
97%
C10H8BrNO2
3-(2-Bromo-5-methoxyphenyl)-3-oxopropanenitrile

Specifications

175136-65-9
25g
C10H8BrNO2
3-(2-Bromo-5-methoxyphenyl)-3-oxopropanenitrile
Amber glass bottle
97%
254.08
97%

Safety and Handling

GHS Signal Word: Warning