Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-[2-(Benzoyloxy)phenyl]propionic acid, 97%

$73.08 - $919.12

Chemical Identifiers

Chemical Name or Material 3-[2-(Benzoyloxy)phenyl]propionic acid
CAS 59725-59-6
Molecular Formula C16H14O4
Formula Weight 270.29
MDL Number MFCD00052018
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2490903

  SDS
alfa aesar™
B2490903
1g Each for $73.08

AAB2490906

  SDS
alfa aesar™
B2490906
5g Each for $260.68

AAB2490914

  SDS
alfa aesar™
B2490914
25g Each for $919.12
Description & Specifications

Specifications

Formula Weight 270.29
Chemical Name or Material 3-[2-(Benzoyloxy)phenyl]propionic acid
Assay 97%
Molecular Formula C16H14O4
MDL Number MFCD00052018
Quantity 1g
CAS 59725-59-6
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.