missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(2-Aminoethyl)-5-bromoindole, 97%

$91.91 - $1,215.19

Chemical Identifiers

CAS 3610-42-2
Molecular Formula C10H11BrN2
Molecular Weight (g/mol) 239.116
MDL Number MFCD00130169
InChI Key CGHUQJRRADEHTQ-UHFFFAOYSA-N
Synonym 5-bromotryptamine, 2-5-bromo-1h-indol-3-yl ethanamine, 5-bromo-1h-indole-3-ethylamine, 1h-indole-3-ethanamine,5-bromo, 2-5-bromo-1h-indol-3-yl ethan-1-amine, 1h-indole-3-ethanamine, 5-bromo, 2-5-bromo-1h-indol-3-yl-ethylamine, 5-bromo-tryptamine, 5-br-t, 3-2-aminoethyl-5-bromoindole
PubChem CID 77158
IUPAC Name 2-(5-bromo-1H-indol-3-yl)ethanamine
SMILES C1=CC2=C(C=C1Br)C(=CN2)CCN
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH63653MD View Documents Alfa Aesar™
H63653MD
250mg Each for $91.91
 
AAH6365303 View Documents Alfa Aesar™
H6365303
1g Each for $291.46
 
AAH6365306 View Documents Alfa Aesar™
H6365306
5g Each for $1,215.19
 
Description

Description

Applications
N-Boc-O-benzyl-D-tyrosine is used as a intermediate for pharmaceutical and chemical research.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Chemical Identifiers

3610-42-2
239.116
CGHUQJRRADEHTQ-UHFFFAOYSA-N
77158
C1=CC2=C(C=C1Br)C(=CN2)CCN
C10H11BrN2
MFCD00130169
5-bromotryptamine, 2-5-bromo-1h-indol-3-yl ethanamine, 5-bromo-1h-indole-3-ethylamine, 1h-indole-3-ethanamine,5-bromo, 2-5-bromo-1h-indol-3-yl ethan-1-amine, 1h-indole-3-ethanamine, 5-bromo, 2-5-bromo-1h-indol-3-yl-ethylamine, 5-bromo-tryptamine, 5-br-t, 3-2-aminoethyl-5-bromoindole
2-(5-bromo-1H-indol-3-yl)ethanamine

Specifications

250mg
C10H11BrN2
5-bromotryptamine, 2-5-bromo-1h-indol-3-yl ethanamine, 5-bromo-1h-indole-3-ethylamine, 1h-indole-3-ethanamine,5-bromo, 2-5-bromo-1h-indol-3-yl ethan-1-amine, 1h-indole-3-ethanamine, 5-bromo, 2-5-bromo-1h-indol-3-yl-ethylamine, 5-bromo-tryptamine, 5-br-t, 3-2-aminoethyl-5-bromoindole
C1=CC2=C(C=C1Br)C(=CN2)CCN
239.116
239.11
97%
3610-42-2
MFCD00130169
CGHUQJRRADEHTQ-UHFFFAOYSA-N
2-(5-bromo-1H-indol-3-yl)ethanamine
77158
Solid
3-(2-Aminoethyl)-5-bromoindole
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning