Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(2-Amino-3-pyridyl)acrylic acid, 97%

$269.72 - $1,193.35

Chemical Identifiers

Chemical Name or Material 3-(2-Amino-3-pyridyl)acrylic acid
CAS 773131-96-7
Molecular Formula C8H8N2O2
Formula Weight 164.17
MDL Number MFCD06205691
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5002103

  SDS
alfa aesar™
H5002103
1g Each for $269.72

AAH5002106

  SDS
alfa aesar™
H5002106
5g Each for $1,193.35
Description & Specifications

Specifications

Formula Weight 164.17
MDL Number MFCD06205691
Assay 97%
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Molecular Formula C8H8N2O2
CAS 773131-96-7
Chemical Name or Material 3-(2-Amino-3-pyridyl)acrylic acid
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.