Please login to your online account to display your discounted pricing

3-(2,6-Dichlorophenyl)-5-methylisoxazole-4-carbonyl chloride, Maybridge

Chemical Identifiers

Chemical Name or Material 3-(2, 6-Dichlorophenyl)-5-methylisoxazole-4-carbonyl chloride
Assay 97%
Assay Percent Range 97%
Molecular Formula C11H6Cl3NO2
Formula Weight 290.53
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SB00576EA

 
fisher scientific company
SB00576EA
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 290.53
Quantity 10g
Assay 97%
Chemical Name or Material 3-(2, 6-Dichlorophenyl)-5-methylisoxazole-4-carbonyl chloride
Assay Percent Range 97%
Molecular Formula C11H6Cl3NO2
CAS 4462-55-9