Please login to your online account to display your discounted pricing

3-(2,6-Dichlorophenyl)-5-methylisoxazole-4-carbonyl chloride, Maybridge


Quantity 10g

Manufacturer: fisher scientific company  SB00576EA

 View more versions of this product

Catalog No. SB00576EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 290.53
Quantity 10g
Assay 97%
Molecular Formula C11H6Cl3NO2
Assay Percent Range 97%
Chemical Name or Material 3-(2, 6-Dichlorophenyl)-5-methylisoxazole-4-carbonyl chloride
CAS 4462-55-9

Specifications

Chemical Name or Material 3-(2, 6-Dichlorophenyl)-5-methylisoxazole-4-carbonyl chloride
Assay 97%
Assay Percent Range 97%
Molecular Formula C11H6Cl3NO2
Formula Weight 290.53