Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(2,6-Dichlorophenoxy)-1-propyne, 97%

$91.46 - $337.51

Chemical Identifiers

Chemical Name or Material 3-(2,6-Dichlorophenoxy)-1-propyne
CAS 3598-66-1
Molecular Formula C9H6Cl2O
Formula Weight 201.05
MDL Number MFCD00052717
Beilstein 2091839
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1267803

  SDS
alfa aesar™
L1267803
1g Each for $91.46

AAL1267806

  SDS
alfa aesar™
L1267806
5g Each for $337.51
Description & Specifications

Specifications

Formula Weight 201.05
Chemical Name or Material 3-(2,6-Dichlorophenoxy)-1-propyne
Assay 97%
CAS 3598-66-1
Molecular Formula C9H6Cl2O
MDL Number MFCD00052717
Beilstein 2091839
Quantity 1g
Odor Odorless
Melting Point 42° to 45°C