missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(2,6-Dichlorophenoxy)-1-propyne, 97%

Manufacturer:  Alfa Aesar™ L1267803

 View more versions of this product

Catalog No. AAL1267803



Specifications

Specifications

3-(2,6-Dichlorophenoxy)-1-propyne
42°C to 45°C
3598-66-1
MFCD00052717
3-2,6-dichlorophenoxy-1-propyne, 1,3-dichloro-2-prop-2-yn-1-yloxy benzene, 1,3-dichloro-2-prop-2-ynyloxy benzene, 1,3-dichloro-2-prop-2-ynyloxybenzene, acmc-20anlz, maybridge1_002901, 2,6-dichlorphenyl-propargylether, 3-2,6-dichlorophenoxy prop-1-yne, 2,6-dichlorophenyl prop-2-yn-1-yl ether, 31401,3-dichloro-2-prop-2-ynyloxy benzene
C#CCOC1=C(C=CC=C1Cl)Cl
201.046
201.05
Odorless
1g
C9H6Cl2O
2091839
HCGOBPUSIBTNRV-UHFFFAOYSA-N
1,3-dichloro-2-prop-2-ynoxybenzene
2776880
97%

Chemical Identifiers

3598-66-1
201.046
HCGOBPUSIBTNRV-UHFFFAOYSA-N
2776880
C#CCOC1=C(C=CC=C1Cl)Cl
C9H6Cl2O
MFCD00052717
3-2,6-dichlorophenoxy-1-propyne, 1,3-dichloro-2-prop-2-yn-1-yloxy benzene, 1,3-dichloro-2-prop-2-ynyloxy benzene, 1,3-dichloro-2-prop-2-ynyloxybenzene, acmc-20anlz, maybridge1_002901, 2,6-dichlorphenyl-propargylether, 3-2,6-dichlorophenoxy prop-1-yne, 2,6-dichlorophenyl prop-2-yn-1-yl ether, 31401,3-dichloro-2-prop-2-ynyloxy benzene
1,3-dichloro-2-prop-2-ynoxybenzene
Documents