Please login to your online account to display your discounted pricing

Alfa Aesar™ 3(2),6,6-Trimethylfulvene, mixture of isomers, tech. 90%

Chemical Identifiers

Chemical Name or Material 3(2),6,6-Trimethylfulvene
Name Note mixture of isomers
Purity Grade Technical Grade
Molecular Formula C6H12
Formula Weight 120.19
MDL Number MFCD02683073
UN Number UN3295
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2182603

  SDS
alfa aesar™
B2182603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2182606

  SDS
alfa aesar™
B2182606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 120.19
Chemical Name or Material 3(2),6,6-Trimethylfulvene
Name Note mixture of isomers
Purity Grade Technical Grade
Assay 90%
Molecular Formula C6H12
MDL Number MFCD02683073
UN Number UN3295
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226
Flammable liquid and vapour.
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226
Flammable liquid and vapour.