Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(2,5-Dimethoxyphenyl)-1H-pyrazole

$123.41 - $123.41

Chemical Identifiers

Chemical Name or Material 3-(2,5-Dimethoxyphenyl)-1H-pyrazole
CAS 181122-45-2
Molecular Formula C11H12N2O2
Formula Weight 204.23
MDL Number MFCD02091528
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5063603

  SDS
alfa aesar™
H5063603
1g Each for $123.41
Description & Specifications

Specifications

Formula Weight 204.23
MDL Number MFCD02091528
CAS 181122-45-2
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
Molecular Formula C11H12N2O2
Chemical Name or Material 3-(2,5-Dimethoxyphenyl)-1H-pyrazole
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.