Please login to your online account to display your discounted pricing

3-(2,4-Dichlorophenyl)propanoic acid, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 3-(2, 4-Dichlorophenyl)propanoic acid
CAS 55144-92-8
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H8Cl2O2
Linear Formula Cl2C6H3CH2CH2CO2H
Formula Weight 219.07
MDL Number MFCD00275605
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SEW04678DA

 
fisher scientific company
SEW04678DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 219.07
Quantity 1g
Assay 97%
CAS Min % 97.0
Chemical Name or Material 3-(2, 4-Dichlorophenyl)propanoic acid
Assay Percent Range 97%
Linear Formula Cl2C6H3CH2CH2CO2H
CAS 55144-92-8
Molecular Formula C9H8Cl2O2
MDL Number MFCD00275605
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Packaging Amber Glass Bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.