Please login to your online account to display your discounted pricing

3-[2-(2-Iodoacetamido)acetamido]-PROXYL, 98%, free radical, ACROS Organics™


Quantity 10mg

Manufacturer: thermo fisher scientific  225000100

 View more versions of this product

Catalog No. AC225000100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Assay ≥97.5%
Formula Weight 382.21
Quantity 10mg
Color Light yellow
TSCA TSCA
CAS Max % 100.0
Health Hazard 1 LIGHT SENSITIVE
Chemical Name or Material 3-[2-(2-Iodoacetamido)acetamido]-PROXYL
Infrared Spectrum Authentic
Name Note 98%
Assay Percent Range 98%
Molecular Formula C12 H21 I N3 O3
MDL Number MFCD00010162
CAS 74648-17-2
Synonym 3-[2-(2-Iodoacetamido)acetamido]-2,2,5,5-tetramethyl-1-pyrrolidinyloxy
CAS Min % 97.5

Specifications

Chemical Name or Material 3-[2-(2-Iodoacetamido)acetamido]-PROXYL
Name Note 98%
CAS 74648-17-2
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C12 H21 I N3 O3
Formula Weight 382.21
MDL Number MFCD00010162
Synonym 3-[2-(2-Iodoacetamido)acetamido]-2,2,5,5-tetramethyl-1-pyrrolidinyloxy
Accessories

LIGHT SENSITIVE

SDS & Safety Documents