Please login to your online account to display your discounted pricing

3-[2-(2-Iodoacetamido)acetamido]-PROXYL, 98%, free radical, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-[2-(2-Iodoacetamido)acetamido]-PROXYL
Name Note 98%
CAS 74648-17-2
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C12 H21 I N3 O3
Formula Weight 382.21
MDL Number MFCD00010162
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC225000100

  SDS
thermo fisher scientific
225000100
10mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 382.21
Physical Form Crystalline Powder
Assay ≥97.5%
Quantity 10mg
Color Light yellow
TSCA TSCA
CAS Max % 100.0
Health Hazard 1 LIGHT SENSITIVE
CAS Min % 97.5
Infrared Spectrum Authentic
Synonym 3-[2-(2-Iodoacetamido)acetamido]-2,2,5,5-tetramethyl-1-pyrrolidinyloxy
MDL Number MFCD00010162
Assay Percent Range 98%
Name Note 98%
Molecular Formula C12 H21 I N3 O3
Chemical Name or Material 3-[2-(2-Iodoacetamido)acetamido]-PROXYL
CAS 74648-17-2

Polymerization reagent

Safety and Handling

LIGHT SENSITIVE