Please login to your online account to display your discounted pricing

3-[2-(2-Bromoacetamido)acetamide]-PROXYL, 98%, free radical, ACROS Organics™


Quantity 10mg

Manufacturer: thermo fisher scientific  224790100

 View more versions of this product

Catalog No. AC224790100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 335.21
Quantity 10mg
Color Cream
TSCA TSCA
CAS Max % 100.0
Chemical Name or Material 3-[2-(2-Bromoacetamido)acetamide]-PROXYL
MDL Number MFCD00010073
CAS 100900-13-8
CAS Min % 97.5
Name Note 98%
Assay Percent Range 98%
Molecular Formula C12H21BrN3O3
Synonym 3-[2-(2-Bromoacetamido)acetamido]-2,2,5,5-tetramethyl- 1-pyrrolidinoxy
Health Hazard 1 LIGHT SENSITIVE
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 3-[2-(2-Bromoacetamido)acetamide]-PROXYL
Name Note 98%
CAS 100900-13-8
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C12H21BrN3O3
Formula Weight 335.21
MDL Number MFCD00010073
Synonym 3-[2-(2-Bromoacetamido)acetamido]-2,2,5,5-tetramethyl- 1-pyrrolidinoxy
Accessories

LIGHT SENSITIVE

SDS & Safety Documents