Please login to your online account to display your discounted pricing

3-[2-(2-Bromoacetamido)acetamide]-PROXYL, 98%, free radical, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-[2-(2-Bromoacetamido)acetamide]-PROXYL
Name Note 98%
CAS 100900-13-8
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C12H21BrN3O3
Formula Weight 335.21
MDL Number MFCD00010073
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC224790250

  SDS
thermo fisher scientific
224790250
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC224790100

  SDS
thermo fisher scientific
224790100
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder
Assay ≥97.5%
Formula Weight 335.21
Quantity 25mg
Color Cream
TSCA TSCA
CAS Max % 100.0
Chemical Name or Material 3-[2-(2-Bromoacetamido)acetamide]-PROXYL
Name Note 98%
Assay Percent Range 98%
Molecular Formula C12H21BrN3O3
MDL Number MFCD00010073
CAS 100900-13-8
Synonym 3-[2-(2-Bromoacetamido)acetamido]-2,2,5,5-tetramethyl- 1-pyrrolidinoxy
CAS Min % 97.5
Health Hazard 1 LIGHT SENSITIVE
Infrared Spectrum Authentic

Polymerization reagent

Safety and Handling

LIGHT SENSITIVE